Please login to your online account to display your discounted pricing

Promega™ Wizard™ SV Gel and PCR Cleanup System DFS Item


Extract DNA fragments from standard or low-melt agarose gels or purify PCR products directly from an amplification reaction. Purified DNA can be used for automated fluorescent DNA sequencing, cloning, labeling, restriction enzyme digestion, or in vitro transcription/translation.

Manufacturer: promega™  A9282

 View more versions of this product

Vacuum protocol requires vacuum adapters (A1331, sold separately).

Catalog No. PR-A9282

  • / Each
 

Description & Specifications

Specifications

Format Spin or Vacuum
Quantity 250 Preps
Product Type System
Includes 100mL Membrane binding solution, 75mL membrane wash solution, 13mL Nuclease-free Water, 250 Wizard™ SV minicolumns, 250 2mL collection tubes
For Use With (Application) Purifies DNA from agarose gels or PCR reactions in as little as 15 min.

Membrane-based system purifies DNA fragments from standard or low-melt agarose gels in either Tris acetate (TAE) or Tris borate (TBE). Also purifies products directly from PCR reactions, removing excess nucleotides, and primers.

  • Effectively extracts DNA ranging from 100bp to 10kb
  • Recovers up to 95% of fragments or amplification products in as little as 15μL elution volume
  • Suitable for spin or vacuum purification formats; no phenol extraction or ethanol precipitation required
  • Tested to ensure performance of purified products in automated fluorescent sequencing to at least 700 bp with 78% accuracy