Please login to your online account to display your discounted pricing

Promega™ Wizard™ SV 96 PCR Cleanup System


Isolates 100 bp to 10kb PCR products directly from a reaction mix

Manufacturer: promega™  A9341

 View more versions of this product

Requires the Vac-Man 96 Vacuum Manifold for manual protocols; automated protocols require a vacuum manifold compatible with the automated system

Catalog No. PR-A9341

  • / Each
 

Description & Specifications

Specifications

Quantity 4 x 96 Preps
Format 96-well
Product Type System
Includes 100mL Membrane binding solution, 150mL Nuclease-free Water, 4 binding plates, 4 collection plate, 1 protocol
For Use With (Application) High-throughput purification of 100bp to 10kb PCR products from excess Nucleotides, primers and primer dimers