Please login to your online account to display your discounted pricing

Promega™ Wizard™ SV 96 PCR Cleanup System DFS Item


Isolates 100 bp to 10kb PCR products directly from a reaction mix

Manufacturer: promega™  A9340

 View more versions of this product

Requires the Vac-Man 96 Vacuum Manifold for manual protocols; automated protocols require a vacuum manifold compatible with the automated system

Catalog No. PR-A9340

  • / Each
 

Description & Specifications

Specifications

Product Type System
For Use With (Application) High-throughput purification of 100bp to 10kb PCR products from excess Nucleotides, primers and primer dimers
Format 96-well
Quantity 96 Preps
Includes 20mL Membrane binding solution, 25mL Nuclease-free Water, 1 binding plate, 1 collection plate, 1 protocol