Please login to your online account to display your discounted pricing
Promega™ Wizard™ PCR Preps DNA Purification System DFS Item
Resin-based system purifies double-stranded PCR-amplified DNA using a 15-minute batch column purification method
$126.95 - $518.98
Specifications
For Use With (Application) Simple, reliable way to purify double-stranded PCR-amplified
View More Specs
Includes:  Wizard SV Purification Resin, Direct Purification Buffer, Wizard Minicolumns, 3cc syringe barrels
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    
PR-A7170
DFS Item
promega™
A7170
50mL Wizard™ PCR preps DNA purification Resin, 5mL direct purification Buffer, 50 Wizard™ minicolumns, 50 3cc syringe barrels, protocol 50 Preps Each for $126.95
PR-A2180
DFS Item
promega™
A2180
250mL Wizard™ PCR preps DNA purification Resin, 25mL direct purification Buffer, 250 Wizard™ minicolumns, 250 3cc syringe barrels, protocol 250 Preps Each for $518.98
Description & Specifications
Specifications
For Use With (Application) Simple, reliable way to purify double-stranded PCR-amplified
  • Recovers 70–90% of DNA fragments from low-melting agarose gels and 95–99% of products from PCR reactions
  • Does not bind fragments smaller than 200 bp, so primers and primer-dimers are removed from the reaction
  • Elutes DNA from the DNA Purification Resin in water or TE buffer, free of salts or macro molecular contaminants
  • Recommended for DNA fragments between 200 and 3200 bp
  • Can easily process multiple PCR Preps at one time with the Vac-Man™ Laboratory Vacuum Manifold (Mfr. No. A7231)