Please login to your online account to display your discounted pricing
Promega™ Wizard™ PCR Preps DNA Purification System DFS Item
Resin-based system purifies double-stranded PCR-amplified DNA using a 15-minute batch column purification method
Manufacturer: promega™  A2180
 View more versions of this product
Catalog No. PR-A2180
  • / Each
Description & Specifications
Specifications
Includes 250mL Wizard™ PCR preps DNA purification Resin, 25mL direct purification Buffer, 250 Wizard™ minicolumns, 250 3cc syringe barrels, protocol
For Use With (Application) Simple, reliable way to purify double-stranded PCR-amplified
Quantity 250 Preps
Product Type System