Please login to your online account to display your discounted pricing

Promega™ Wizard™ Plus SV Minipreps DNA Purification System


Produces high yield plasmid DNA of consistent quality from bacterial cultures

Manufacturer: promega™  A1340

 View more versions of this product

 Disclaimers

U.S. Patent 5,981,235, Australian Patent 729932.

Catalog No. PR-A1340

  • / Each
 

Description & Specifications

Specifications

Yield 1 to 20μg
Quantity 1-10mL
Storage Requirements Store at 22° to 25°C.
For Use With (Application) Produces high yield plasmid DNA of consistent quality from bacterial cultures
Product Type DNA Purification System plus Vacuum Adapters
Includes 50 Wizard SV minicolumns, 50 Collection tubes, 20mL Column wash solution, 550μL Alkaline Protease solution, 30mL Neutralization solution, 20mL Cell resuspension solution, 20mL Cell Lysis solution, 13mL Nuclease-free water, 20 Vacuum adapters
No. of Reactions 50 Preps
Format Spin or vacuum