Please login to your online account to display your discounted pricing

Promega™ Wizard™ Plus SV Minipreps DNA Purification System DFS Item


Produces high yield plasmid DNA of consistent quality from bacterial cultures

Manufacturer: promega™  A1330

 View more versions of this product

 Disclaimers

U.S. Patent 5,981,235, Australian Patent 729932.

Catalog No. PR-A1330

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at 22° to 25°C.
Yield 1 to 20μg
Format Spin or vacuum
Includes 50 Wizard™ SV minicolumns, 50 collection tubes, 20mL column wash solution, 550μL Alkaline Protease solution, 30mL neutralization solution, 20mL Cell resuspension solution, 20mL Cell Lysis solution, 13mL Nuclease-free Water
For Use With (Application) Produces high yield plasmid DNA of consistent quality from bacterial cultures
No. of Reactions 50 Preps
Quantity 1-10mL
Product Type DNA Purification System