Please login to your online account to display your discounted pricing

Promega™ Wizard™ Plus DNA Purification Systems


Purify plasmid DNA from culture at the mini-, midi-, maxi-, or megaprep scale

Manufacturer: promega™  A7510

 View more versions of this product

Catalog No. PR-A7510

  • / Each
 

Description & Specifications

Specifications

Format Manual
Includes 50mL Cell resuspension solution, 50mL Cell Lysis solution, 100mL neutralization solution, 250 Wizard™ minicolumns, 250mL Wizard™ Minipreps DNA purification Resin, 250 3mL syringe barrels, 230mL column wash solution
Quantity 1-3mL
Yield 1 to 10μg
No. of Reactions 250 Preps
For Use With (Application) Purifies plasmid DNA from culture
Product Type Kit