Please login to your online account to display your discounted pricing

Promega™ Wizard™ Plus DNA Purification Systems


Purify plasmid DNA from culture at the mini-, midi-, maxi-, or megaprep scale

Manufacturer: promega™  A7500

 View more versions of this product

Catalog No. PR-A7500

  • / Each
 

Description & Specifications

Specifications

Format Manual
Yield 1 to 10μg
For Use With (Application) Purifies plasmid DNA from culture
Quantity 1-3mL
Product Type Kit
No. of Reactions 100 Preps
Includes 20mL Cell resuspension solution, 20mL Cell Lysis solution, 40mL neutralization solution, 100 Wizard™ minicolumns, 100mL Wizard™ Minipreps DNA purification Resin, 100 3mL syringe barrels, 100mL column wash solution