Please login to your online account to display your discounted pricing

Promega™ Wizard™ Plus DNA Purification Systems DFS Item


Purify plasmid DNA from culture at the mini-, midi-, maxi-, or megaprep scale

Manufacturer: promega™  A7270

 View more versions of this product

Catalog No. PR-A7270

  • / Each
 

Description & Specifications

Specifications

Format Manual
Quantity 100-500mL
Includes 150mL Cell resuspension solution, 150mL Cell Lysis solution, 300mL neutralization solution, 100mL Wizard™ Maxipreps DNA purification Resin, 125mL Column wash solution, 10 maxi/megacolumns with reservoirs, 10 5mL syringes, 10 0.2
Yield 300 to 1000μg
No. of Reactions 10 Preps
For Use With (Application) Purifies plasmid DNA from culture
Product Type Kit