Please login to your online account to display your discounted pricing

Promega™ Wizard™ DNA Purification Resins


Resins for use with Wizard DNA purification kits

Manufacturer: promega™  A7141

 View more versions of this product

Catalog No. PR-A7141

  • / Each
 

Description & Specifications

Specifications

Format Liquid
For Use With (Application) Rapid isolation of plasmid DNA
Quantity 250mL
For Use With (Equipment) Wizard™ Plus Minipreps DNA purification systems
Size 250mL
Product Type Minipreps Resin
Product Name Minipreps Resin
Quantity 250mL