Please login to your online account to display your discounted pricing

Promega™ Wizard™ DNA Purification Resins DFS Item


Resins for use with Wizard DNA purification kits

Manufacturer: promega™  A7181

 View more versions of this product

Catalog No. PR-A7181

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Reliable way to purify double-stranded PCR-amplified DNA
For Use With (Equipment) Wizard™ PCR Preps DNA Purification System
Quantity 250mL
Format Liquid
Product Type PCR Preps Resin