Please login to your online account to display your discounted pricing

Promega™ Wizard™ DNA Purification Resins


Resins for use with Wizard DNA purification kits

Manufacturer: promega™  A7701

 View more versions of this product

Catalog No. PR-A7701

  • / Each
 

Description & Specifications

Specifications

Product Type Midipreps Resin
For Use With (Application) Rapid isolation of plasmid DNA
Format Liquid
Size 1000mL
Product Name Midipreps Resin
Quantity 1000mL
Quantity 1000mL
For Use With (Equipment) Wizard™ Plus Midipreps DNA Purification System