missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ Ultra Pure Deoxynucleotide Triphosphates (dNTPs) Sets DFS Item

High-quality dNTPs. Provided at a concentration of 100mM in water at pH 7.5.

Manufacturer:  Promega™ U1410

 View more versions of this product

Catalog No. PR-U1410



Description

Description

  • Triphosphate content is >98%
  • No detectable DNase or RNase activity
  • Performance is tested using AMV reverse transcriptase, DNA polymerase, and Taq
  • DNA polymerase dNTPs are assayed to ensure the absence of contaminating dUTP
  • Includes individual tubes of each dNTP

Specifications

Specifications

100mM
Individual tubes of each dNTP
>99%
-30° to -10°C
cDNA synthesis, sequencing and labeling, can be used in place of dTTP in PCR and RT-PCR protocols
7.5
4 x (2 x 100μmol)
Documents
Promotions

Promotions