Please login to your online account to display your discounted pricing

Promega™ Ultra Pure Deoxynucleotide Triphosphates (dNTPs): PCR Nucleotide Mix


Contains equal amounts of each dNTP (dATP, dCTP, dGTP and dTTP) in a single tube

Manufacturer: promega™  C1145

 View more versions of this product

Catalog No. PR-C1145

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) cDNA synthesis, sequencing and labeling, can be used in place of dTTP in PCR and RT-PCR protocols
pH 7.5
Concentration 10mM
Purity >98%
Quantity 1000μL
Storage Requirements -20°C