Promega™ UGT-Glo™ Activity Assays DFS Item
Uses a luminescent method to measure UGT activity
Manufacturer:  Promega™ V2081
 View more versions of this product
Catalog No. PRV2081
Specifications Description & Specifications
Specifications
UGT-Glo assay
-70°C
UGT multienzyme Substrate, UGT1A4 Substrate, Reconstitution Buffer, Luciferin detection reagent, 5X UGT Buffer, 50mM UDPGA, 100X D-Cysteine, proluciferin Substrates
200 Assays
Luminescent method to measure UGT activity
Promotions
Promotions
  • Purchase up to $1,000 in any combination of Promega™ reagents and receive an equivalent amount of complimentary Promega products of your choice. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.
    06/30/18