Promega UGT-Glo™ Activity Assays DFS Item
The UGT-Glo™ Assay provides a luminescent method for measuring UDP glucuronosyltransferase (UGT) activity. The UGT-Glo™ Assay is designed to measure UGT activity from a variety of sources and to test the effects of various chemicals on UGT activity.
Manufacturer:  Promega V2082
 View more versions of this product
Catalog No. PRV2082
Description
Description
  • Includes:UGT Multienzyme Substrate, UGT1A4 Substrate, Reconstitution Buffer, Luciferin Detection Reagent, UGT Buffer, 5X UDPGA, 50mM D-Cysteine, 100X
Specifications
Specifications
Luminescent method to measure UGT activity
UGT-Glo assay
-70°C
UGT multienzyme Substrate, UGT1A4 Substrate, Reconstitution Buffer, Luciferin detection reagent, 5X UGT Buffer, 50mM UDPGA, 100X D-Cysteine, proluciferin Substrates
1000 Assays