missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega UGT-Glo™ Activity Assays DFS Item

The UGT-Glo™ Assay provides a luminescent method for measuring UDP glucuronosyltransferase (UGT) activity. The UGT-Glo™ Assay is designed to measure UGT activity from a variety of sources and to test the effects of various chemicals on UGT activity.

$1,651.02 - $1,651.02

Specifications

Product Type UGT-Glo assay
For Use With (Application) Luminescent method to measure UGT activity
Includes UGT multienzyme Substrate, UGT1A4 Substrate, Reconstitution Buffer, Luciferin detection reagent, 5X UGT Buffer, 50mM UDPGA, 100X D-Cysteine, proluciferin Substrates
Storage Requirements -70°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRV2082 View Documents Promega
V2082
1000 Assays Each for $1,651.02
 
Description

Description

  • A Luminescent Method for Measuring UGT Activity from a Variety of Sources
  • Luminescent format eliminates need for time-consuming analyses
  • Simple and sensitive protocol saves time and money
  • UGT2B7 Screening System is no longer available
Specifications

Specifications

UGT-Glo assay
Luminescent method to measure UGT activity
UGT multienzyme Substrate, UGT1A4 Substrate, Reconstitution Buffer, Luciferin detection reagent, 5X UGT Buffer, 50mM UDPGA, 100X D-Cysteine, proluciferin Substrates
-70°C