missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega TRKA Kinase Enzyme System DFS Item

Recombinant human TRKA (amino acids 440-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. TRKA is a member of the trk proto-oncogene family and encodes a 140kDa membrane-spanning protein tyrosine kinase.

$1,230.58 - $1,230.58

Specifications

Storage Requirements COMPONENTS AT DIFFERENT TEMPS
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRV9761 View Documents Promega™
V9761
1/ea Each for $1,230.58
 
Description

Description

  • Easily Screen and Profile TRKA Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications

Specifications

COMPONENTS AT DIFFERENT TEMPS