Please login to your online account to display your discounted pricing

Promega™ Transcription Factor Consensus Oligonucleotides DFS Item


Contain consensus DNA-binding sites for individual sequence-specific transcription factors

Manufacturer: promega™  E3292

 View more versions of this product

Catalog No. PR-E3292

  • / Each
 

Description & Specifications

Specifications

Quantity 35pM
Product Type Oligonucleotide, NF-κB
For Use With (Application) Affinity chromatography, Gel shift assays
Concentration 1.75pM/μL