Please login to your online account to display your discounted pricing

Promega™ Transcription Factor Consensus Oligonucleotides


Contain consensus DNA-binding sites for individual sequence-specific transcription factors

Manufacturer: promega™  E3291

 View more versions of this product

Catalog No. PR-E3291

  • / Each
 

Description & Specifications

Specifications

Concentration 1.75pM/μL
For Use With (Application) Affinity chromatography, Gel shift assays
Product Type Oligonucleotide, NF-κB
Quantity 175pM
Product Name Oligonucleotide, NF-κB
Quantity 175pM
Concentration 1.75pM/μL