Please login to your online account to display your discounted pricing

Promega™ Transcription Factor Consensus Oligonucleotides DFS Item

Contain consensus DNA-binding sites for individual sequence-specific transcription factors

$42.80 - $99.51

Specifications

For Use With (Application) Affinity chromatography, Gel shift assays
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

PR-E3291

DFS Item
 
promega™
E3291
175pM Each for $99.51

PR-E3292

DFS Item
 
promega™
E3292
35pM Each for $42.80
Description & Specifications

Specifications

For Use With (Application) Affinity chromatography, Gel shift assays

  • Double-stranded oligonucleotides designed with 5' OH blunt ends, making them easily labeled to high specific activity with T4 polynucleotide kinase
  • Concentration 1.75pmol/μL

Gel shift assays; Affinity chromatography