Please login to your online account to display your discounted pricing
Promega™ Transcend™ Nonradioactive Translation Detection Systems DFS Item
Allow nonradioactive detection of proteins synthesized in vitro
$323.14 - $472.94
Specifications
For Use With (Application) Non-radioactive detection of Proteins synthesized in-vitro
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    
PR-L5070
DFS Item
promega™
L5070
30μL Transcend™ Trna, 36μL Streptavidin-AP, 35mL Western Blue™ stabilized substrate for Alkaline Phosphatase Each for $323.14
PR-L5080
DFS Item
promega™
L5080
30μL Transcend™ tRna, 0.25mg/mL 15μL Streptavidin-HRP, 25mL Transcend™ Chemiluminescent substrate A and B Each for $472.94
Description & Specifications
Specifications
For Use With (Application) Non-radioactive detection of Proteins synthesized in-vitro
Using these systems, biotinylated lysine residues are incorporated into nascent proteins during translation, eliminating the need for labeling with [35S]methionine or other radioactive amino acids. This biotinylated lysine is added to the translation reaction as a precharged ε-labeled biotinylated lysine-tRNA complex (Transcend tRNA) rather than a free amino acid. After SDS-PAGE and electroblotting, the biotinylated proteins can be visualized by binding either Streptavidin-Alkaline Phosphatase (Streptavidin-AP) or Streptavidin-Horseradish Peroxidase (Streptavidin-HRP), followed either by colorimetric or chemiluminescent detection.
  • Sensitive: The biotin tag allows detection of 0.5 to 5ng of translated protein
  • Safe: No radioisotope handling, storage or disposal is required
  • Fast: Labeled proteins can be detected three to four hours after gel electrophoresis
  • Flexible: Results can be visualized by using colorimetric or chemiluminescent detection
  • IRNA available separately (PR-L5061)