Please login to your online account to display your discounted pricing
Promega™ Transcend™ Nonradioactive Translation Detection Systems DFS Item
Allow nonradioactive detection of proteins synthesized in vitro
Manufacturer: promega™  L5080
 View more versions of this product
Catalog No. PR-L5080
  • / Each
Description & Specifications
Specifications
Quantity 30 x 50μL Reactions
Product Type Transcend Chemiluminescent Nonradioactive Translation Detection System
Includes 30μL Transcend™ tRna, 0.25mg/mL 15μL Streptavidin-HRP, 25mL Transcend™ Chemiluminescent substrate A and B
For Use With (Application) Non-radioactive detection of Proteins synthesized in-vitro