Promega™ ToxGlo™ Mitochondrial Toxicity Assay DFS Item
The Mitochondrial ToxGlo™ Assay provides a method for predicting potential mitochondrial dysfunction as a result of xenobiotic exposure.
Manufacturer:  Promega™ G8000
 View more versions of this product
Catalog No. PRG8000
Description
Description
  • Predictive for mitochondrial toxicity
  • Results in under an hour
  • Easy automation
  • Simple detection with a standard plate reader
Specifications
Specifications
10mL
7.5
<1 Hour
Fluid
1 x 10μL bis-AAF-R110 substrate, 1 x 10mL Assay Buffer, 1 x 10mL ATP detection Buffer, ATP detection substrate
−30 to −10°C