Promega TNT™ SP6 High-Yield Protein Expression System
TNT SP6 High-Yield Wheat Germ Protein Expression System, based on an optimized wheat germ extract, is a single-tube, coupled transcription/translation system designed to express proteins in only two hours.
Supplier: Promega L3260
Includes: TNT SP6 High-Yield Master Mix, Nuclease-free water
Description
- Coupled Transcription/Translation for Soluble, Full-Length Protein in Only Two Hours
- Express soluble, full-length protein from plasmid or PCR-generated templates quickly
- Generate 10- to 20-fold more protein (10-100μg/ml) than with other cell-free systems
Specifications
TnT™ SP6 high-yield master mix, Nuclease-free Water | |
10 to 100 μg/mL | |
40 Reactions | |
<-65°C |
2 hr. | |
Wheat Germ Protein Expression System | |
40 reactions |
U.S. Patent 5,324,637; Australian Patent 660329 and other patents.
For Research Use. Any use of the product for diagnostics requiring clearance or approval by the FDA may require a license under Mayo Clinic U.S. Patents 6,027,913 and 6,361,949.