Please login to your online account to display your discounted pricing
Promega™ TNT™ SP6 High-Yield Protein Expression System
A single-tube, coupled transcription/translation system designed to express proteins in only two hours
Manufacturer: promega™  L3261
 View more versions of this product
Catalog No. P12326A
  • / Each
Description & Specifications
Specifications
Includes TnT™ SP6 high-yield master mix, Nuclease-free Water
Product Type TnT™ SP6 high yield Wheat Germ Protein Expression system
Quantity 10 Reactions
Storage Requirements -70°C
Test Time 2 hr.
Yield 10 to 100μg/mL
  • Based on an optimized wheat germ extract
  • Contains all components necessary for transcription/translation, with the exception of the plasmid DNA or PCR template and optional protein-labeling reagents
  • Yields 10 to 100μg/mL protein, 10- to 20-fold more protein compared to other cell-free systems
  • Generates soluble, full-length protein and avoids problems associated with Escherichia coli systems
  • Produces protein that can be used in multiple proteomic-based applications, as well as in high-throughput analysis