Promega Thermolysin
Thermolysin is a thermostable metalloproteinase. The high digestion temperatures may be used as an alternative to denaturants to improve digestion of proteolytically resistant proteins. The optimal digestion temperature range is 65-85°C.
Manufacturer: Promega V4001
Description
- Thermostable Proteinase with High Digest Temperature; Better Denaturation, Digestion of Proteolytically Resistant Proteins
- Cleaves at the N-terminus of Leu, Phe, Val, Ile, Ala, Met
- Optimal digestion temperature is 65-85°C
- Optimal activity at pH 5.0-8.5
Specifications
25 mg | |
−30 to −10°C | |
149°F to 149°F | |
65°C to 85°C |
Promotions
-
Get Up to $1,000 in Promega Products with Purchase
Purchase up to $1,000 in Promega reagents and get a matching amount of products at no additional cost.: 06/30/21
-
Spend More, Get More on Promega Products
Purchase up to $1,000 in Promega reagents and get a matching amount of products at no additional cost.: 06/30/21