Please login to your online account to display your discounted pricing

Promega™ Terminal Deoxynucleotidyl Transferase

Enzymes are available in convenient, cost-effective concentrations that are economical and easy to pipet

$91.00 - $318.00

Specifications

Color Colorless
Concentration 30U/μL
Product Name Terminal Deoxynucleotidyl Transferase
Format Liquid
For Use With (Application) Tailing reactions to add complementary Homopolymer tails to DNA vectors and cDNA, 3' end-labeling, TUNEL Assays
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

PR-M1875

 
promega™
M1875
1500U Each for $318.00

PR-M1871

 
promega™
M1871
300U Each for $91.00
Description & Specifications

Specifications

Color Colorless
Concentration 30U/μL
Product Name Terminal Deoxynucleotidyl Transferase
Format Liquid
For Use With (Application) Tailing reactions to add complementary Homopolymer tails to DNA vectors and cDNA, 3' end-labeling, TUNEL Assays
Includes 5X Reaction Buffer: 500mM Cacodylate Buffer (pH 6.8 at 25°C), 5mM CoCl2, 0.5mM DTT
pH 6.4
Source Recombinant E. coli strain
Storage Buffer 50mM Potassium Phosphate (pH 6.4), 100mM NaCl, 1mM beta-mercaptoethanol, 0.1% Tween™ 20, 50% Glycerol
Storage Requirements -20°C