Please login to your online account to display your discounted pricing

Promega™ T4 DNA Ligase DFS Item


Enzymes are available in convenient, cost-effective concentrations that are economical and easy to pipet

Manufacturer: promega™  M1801

 View more versions of this product

Catalog No. PR-M1801

  • / Each
 

Description & Specifications

Specifications

Product Name T4 DNA Ligase
Purity >90%
For Use With (Application) Joining double-stranded DNA molecules with cohesive or blunt ends (5', 3' or blunt-ended DNA inserts)
pH 7.4
Storage Requirements -20°C
Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 50% Glycerol
Source Recombinant E. coli strain
Molecular Weight 68
Color Colorless
Concentration 1 to 3U/μL
Includes 10X Reaction Buffer: 300mM Tris HCl (pH 7.8 at 25°C), 100mM MgCl2, 100mM DTT and 10mM ATP
Quantity 100U
Format Liquid