Please login to your online account to display your discounted pricing

Promega™ StemElite™ Human Pluripotent Transcripts DFS Item

Functionally associated with maintenance of the undifferentiated human embryonic stem cell

$104.86 - $104.86

Specifications

Concentration 20X
Format Liquid
For Use With (Application) Characterize the transcription of markers associated with the state of differentiation
pH 7.4
Quantity 100μL
View More Specs

 Disclaimers

The purchase of this product conveys to the buyer the limited, nonexclusive, nontransferable right (without the right to resell, repackage, or further sublicense) under U.S. Published Patent Appln. 20020150900 and U.S. Patents 5,432,272; 6,617,106 and 6,140,496 to use the product. No other license is granted to the buyer whether expressly, by implication, by estoppel or otherwise. In particular, the purchase of this product does not include or carry any right or license to sell this product. For information on purchasing a license for other uses, please contact Promega Corporation, Business Development, 2800 Woods Hollow Road, Madison, WI 53711, or EraGen Biosciences, Corporate Licensing, 918 Deming Way, Suite 201, Madison, WI 53717. Phone (608) 662-9000; Fax (608) 662-9003.

Products
Catalog Number Mfr. No. Primer Product Type Price Quantity    

PRB1011

DFS Item
 
promega™
B1011
NANOG or GAPDH NANOG or GAPDH Primer Pair Each for $104.86

PRB1041

DFS Item
 
promega™
B1041
LIN28 or GAPDH LIN28 or GAPDH Primer Pair Each for $104.86

PRB1031

DFS Item
 
promega™
B1031
POU5F1 or GAPDH POU5F1/GAPDH Primer Pair Each for $104.86

PRB1021

DFS Item
 
promega™
B1021
SOX2 or GAPDH SOX2 or GAPDH Primer Pair Each for $104.86

PRB1051

DFS Item
 
promega™
B1051
KLF4 or GAPDH KLF4 or GAPDH Primer Pair Each for $104.86

PRB1061

DFS Item
 
promega™
B1061
MYC or GAPDH MYC or GAPDH Primer Pair Each for $104.86
Description & Specifications

Specifications

Concentration 20X
Format Liquid
For Use With (Application) Characterize the transcription of markers associated with the state of differentiation
pH 7.4
Quantity 100μL
Storage Requirements -20°C

Permit the amplification of the transcript of interest and reference transcript (GAPDH) in a single tube.