Please login to your online account to display your discounted pricing

Promega™ StemElite™ Human Pluripotent Transcripts


Functionally associated with maintenance of the undifferentiated human embryonic stem cell

Manufacturer: promega™  B1021

 View more versions of this product

 Disclaimers

The purchase of this product conveys to the buyer the limited, nonexclusive, nontransferable right (without the right to resell, repackage, or further sublicense) under U.S. Published Patent Appln. 20020150900 and U.S. Patents 5,432,272; 6,617,106 and 6,140,496 to use the product. No other license is granted to the buyer whether expressly, by implication, by estoppel or otherwise. In particular, the purchase of this product does not include or carry any right or license to sell this product. For information on purchasing a license for other uses, please contact Promega Corporation, Business Development, 2800 Woods Hollow Road, Madison, WI 53711, or EraGen Biosciences, Corporate Licensing, 918 Deming Way, Suite 201, Madison, WI 53717. Phone (608) 662-9000; Fax (608) 662-9003.

Catalog No. PRB1021

  • / Each
 

Description & Specifications

Specifications

Quantity 100μL
Product Type SOX2 or GAPDH Primer Pair
Format Liquid
Storage Requirements -20°C
pH 7.4
Primer SOX2 or GAPDH
Concentration 20X
For Use With (Application) Characterize the transcription of markers associated with the state of differentiation
Product Name SOX2 or GAPDH Primer Pair
Storage Conditions -20°C