Please login to your online account to display your discounted pricing

Promega™ Steady-Glo™ Luciferase Assay System


Provides long-lived luminescence when added to cultured cells

Manufacturer: promega™  E2520

 View more versions of this product

Catalog No. PR-E2520

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) 96- and 384-well plates
Detection Method Luminescence
Storage Requirements Store Steady-Glo™ Luciferase Assay Buffer below 25°C, Steady-Glo™ Luciferase Assay substrate at -20°C
For Use With (Application) For quantitation of firefly luciferase expression in mammalian cells
Quantity 100mL

  • Ideal for quantitation of firefly luciferase expression in mammalian cells by batch processing of 96- and 384-well plates
  • Can process several thousand samples per hour with high reproducibility under standard laboratory conditions
  • Provides signal half-lives over five hours in commonly used cell culture media without prior sample processing
  • Results in greater assay sensitivity than other extended-lifetime firefly luciferase assay reagents
  • Less sensitive to mixing conditions for improved assay reproducibility
  • Compatible with many tissue culture media, including those containing up to 10% serum