missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ SP6 RNA Polymerase DFS Item

DNA-dependent RNA polymerase for in vitro transcription.

Manufacturer:  Promega™ P1085

 View more versions of this product

Catalog No. PR-P1085



Description

Description

  • Applications: Generation of templates for in-vitro translation, generation of probes for Nucleic Acid hybridization, generation of RNA processing Substrates, generation of Antisense RNA
Specifications

Specifications

SP6 RNA Polymerase
10 to 20U/μL
100mM DTT 5X Reaction Buffer, transcription optimized 5X Buffer: 200mM Tris HCl (pH 7.9 at 25°C), 30mM MgCl2, 10mM Spermidine, 50mM NaCl
>90%
1000U
20mM Potassium Phosphate (pH 7.7 at 25°C), 1mM EDTA, 10mM DTT, 100mM NaCl, 0.1% Triton™ X-100, 50% Glycerol
Liquid
Colorless
Recombinant E. coli strain
7.5
Generation of templates for in-vitro translation, generation of probes for Nucleic Acid hybridization, generation of RNA processing Substrates, generation of Antisense RNA
−30 to −10°C
Documents
Promotions

Promotions