Please login to your online account to display your discounted pricing

Promega™ SIRT-Glo™ Assay and Screening Systems


Single-reagent-addition,homogeneous, luminescent assays that measure the relative activity of the NAD+-dependent histone deacetylase (HDAC) class III enzymes (sirtuins; SIRTs) from purified enzyme sources

Manufacturer: promega™  G6450

 View more versions of this product

Catalog No. PRG6450

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Determining SIRT inhibitor or activator potency, SIRT Enzyme profiling
Storage Requirements -20°C
Type Assay
Quantity 10mL
pH 8
Format Fluid
Kit Contents 1 x 1 Vial SIRT-Glo™ substrate, 1 x 10μL developer reagent, 1 x 30μL Nicotinamide, 1 x 25mL SIRT-Glo™ Buffer