Please login to your online account to display your discounted pricing

Promega™ SIRT-Glo™ Assay and Screening Systems

Single-reagent-addition,homogeneous, luminescent assays that measure the relative activity of the NAD+-dependent histone deacetylase (HDAC) class III enzymes (sirtuins; SIRTs) from purified enzyme sources

$621.68 - $621.68

Specifications

Format Fluid
For Use With (Application) Determining SIRT inhibitor or activator potency, SIRT Enzyme profiling
pH 8
Storage Requirements -20°C
View More Specs
Products
Catalog Number Mfr. No. Kit Contents Quantity Type Price Quantity    

PRG6450

 
promega™
G6450
1 x 1 Vial SIRT-Glo™ substrate, 1 x 10μL developer reagent, 1 x 30μL Nicotinamide, 1 x 25mL SIRT-Glo™ Buffer 10mL Assay Each for $621.68
Description & Specifications

Specifications

Format Fluid
For Use With (Application) Determining SIRT inhibitor or activator potency, SIRT Enzyme profiling
pH 8
Storage Requirements -20°C

  • Assay Advantages: simple measurement of deacetylating activities
  • Highly sensitive, fast data acquisition