Please login to your online account to display your discounted pricing

Promega™ SignaTECT™ Protein Kinase Assay Systems DFS Item


Contain proprietary SAM2 Biotin Capture Membrane for higher binding capacity and specificity

Manufacturer: promega™  V7870

 View more versions of this product

Catalog No. PR-V7870

  • / Each
 

Description & Specifications

Specifications

Product Type DNA-dependent Protein Kinase assay system
Quantity 96 Reactions
Includes ATP, BSA, SAM2 Biotin capture membrane, Termination Buffer, CaM KII Biotinylated Peptide Substrate, reaction control, activation 5X Buffer
Storage Requirements -20°C
For Use With (Application) Can be used in a variety of Buffer or reaction conditions including cell extracts