Please login to your online account to display your discounted pricing

SignaTECT™ Protein Kinase Assay Systems


Contain proprietary SAM2 Biotin Capture Membrane for higher binding capacity and specificity

Manufacturer: promega corportaion  V6430

 View more versions of this product

Catalog No. PR-V6430

  • / Each
 

Description & Specifications

Specifications

Quantity 96 Reactions
Storage Requirements -20°C
Product Type cdc2 Protein Kinase assay system
For Use With (Application) Can be used in a variety of Buffer or reaction conditions including cell extracts