Please login to your online account to display your discounted pricing

Promega™ SAM2™ Biotin Capture Membrane


Rapidly bind biotinylated molecules based on their affinity for streptavidin

Manufacturer: promega™  V7861

 View more versions of this product

Catalog No. PR-V7861

  • / Each
 

Description & Specifications

Specifications

Length (Metric) Exterior 7.6cm
Width (Metric) Exterior 10.9cm
For Use With (Application) Protein kinase activity quantitation, specific mRNA quantitation, cytokine quantitation, and specific gene quantitation

  • Provide quantitative, linear substrate binding in the nmol/cm2 range due to the high density of streptavidin on the membrane, permitting kinetic studies
  • Optimized for low nonspecific binding
  • Can be used to analyze a wide variety of biotinylated substances without optimizing each substrate for binding to a matrix
  • Bind within one minute
  • Provide linear binding of substrates up to the nmol/cm2 range, permitting kinetic studies
  • Ideal for protein kinase activity quantitation, specific mRNA quantitation, cytokine quantitation, and specific gene quantitation

SAM2 Biotin Capture Membrane

  • Available as a prenumbered, partially cut sheet, which permits easy separation into 96 individual squares
  • Also available as an uncut sheet, which allows for sample application using a multichannel pipetter
  • Can be analyzed using phosphorimaging analysis, autoradiography, scintillation counting, or chemiluminescent detection to quantitate results
SAM2 96 Biotin Capture Plate
  • Contains SAM2 Biotin Capture Membrane in the wells of a microfiltration plate
  • Allows washes to be performed using a vacuum manifold or a plate washer
  • Supplied with a transparent top seal and an opaque bottom seal, allowing addition of scintillation fluid for data quantitation using a microplate liquid scintillation counter