Please login to your online account to display your discounted pricing

Promega™ rNTPs DFS Item


Ideal for in vitro transcription

Manufacturer: promega™  E6041

 View more versions of this product

Catalog No. PR-E6041

  • / Each
 

Description & Specifications

Specifications

Quantity 400μL
Storage Requirements -20°C
For Use With (Application) Provides a cell-free system for in-vitro transcription of eukaryotic genes
Concentration 100mM
Product Type rCTP