Please login to your online account to display your discounted pricing

Promega™ Riboprobe rNTPs


Qualified for use with the Riboprobe Systems

Manufacturer: promega™  P1152

 View more versions of this product

Catalog No. PR-P1152

  • / Each
 

Description & Specifications

Specifications

Product Type rGTP
Quantity 0.5mL
For Use With (Application) Examining antimicrobial compound activity, screening antimicrobial compounds, examining bacterial growth with extended sensitivity and range, rapid bacterial quantitation, rapid ATP detection
Concentration 10mM
Storage Requirements -20°C
Purity HPLC
Product Type rGTP