Please login to your online account to display your discounted pricing

Promega™ Riboprobe rNTPs


Qualified for use with the Riboprobe Systems

Manufacturer: promega™  P1132

 View more versions of this product

Catalog No. PR-P1132

  • / Each
 

Description & Specifications

Specifications

Quantity 0.5mL
Storage Requirements -20°C
For Use With (Application) Examining antimicrobial compound activity, screening antimicrobial compounds, examining bacterial growth with extended sensitivity and range, rapid bacterial quantitation, rapid ATP detection
Product Type rATP
Concentration 10mM
Purity HPLC
Product Type rATP