Please login to your online account to display your discounted pricing

Promega™ Riboprobe rNTPs


Qualified for use with the Riboprobe Systems

Manufacturer: promega™  P1221

 View more versions of this product

Catalog No. PR-P1221

  • / Each
 

Description & Specifications

Specifications

Quantity 4 x 0.5mL
Storage Requirements -20°C
Concentration 10mM
Product Type rNTP Set (rATP, rCTP, rGTP, rUTP)
For Use With (Application) Examining antimicrobial compound activity, screening antimicrobial compounds, examining bacterial growth with extended sensitivity and range, rapid bacterial quantitation, rapid ATP detection
Purity HPLC