Promega™ Restriction Enzymes - Taq I (E) DFS Item
Sensitive to overlapping dam methylation. (T/CG A A GC\T)
Manufacturer:  Promega™ R4154
 View more versions of this product
Catalog No. PR-R4154
Specifications
Specifications
TaqI (HC)
40 to 80U/μL
1 x 1mL 10X Buffer E, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 5000U TaqI (HC), 1 x 0.25mL Multi-Core™ 10X Buffer
7.4
10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Liquid
T.CGA AGC.T
37°C
5,000U
-20°C
Documents