Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Xho I Blue/White Cloning and Genome Qualified (D)


Manufacturer: promega™  R4164

 View more versions of this product

Catalog No. PR-R4164

  • / Each
 

Description & Specifications

Specifications

Product Name XhoI (HC)
Quantity 15000U
For Use With (Application) Provides a higher level of quality control for Enzymes used in cloning applications
Storage Buffer 10mM Tris HCl (pH 7.3), 300mM KCl, 0.1% Triton™ X-100, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Concentration 40 to 80U/μL
Units 15,000U
pH 7.3
Product Name XhoI (HC)
Cut Site C.TCGAG GAGCT.C
Includes 1 x 1mL 10X Buffer D, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 15000U XhoI (HC)
Format Liquid
Incubator Temperature 37°C
Format Liquid
Applications Provides a higher level of quality control for Enzymes used in cloning applications
Quantity 15000U