Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Xba I Blue/White Cloning and Genome Qualified (D)


Manufacturer: promega™  R4184

 View more versions of this product

Catalog No. PR-R4184

  • / Each
 

Description & Specifications

Specifications

Product Name XbaI (HC)
Cut Site T.CTAGA AGATC.T
Quantity 10000U
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Incubator Temperature 37°C
pH 7.4
Format Liquid
For Use With (Application) Provides a higher level of quality control for Enzymes used in cloning applications
Includes 1 x 1mL 10X Buffer D, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 10000U XbaI (HC), 1 x 0.25mL Multi-Core™ 10X Buffer
Concentration 40 to 80U/μL
Units 10,000U