Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Ssp I Genome Qualified (E)


Manufacturer: promega™  R6601

 View more versions of this product

Catalog No. PR-R6601

  • / Each
 

Description & Specifications

Specifications

Product Name SspI
Incubator Temperature 37°C
Includes 1 x 1mL 10X Buffer E, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U SspI, 1 x 0.25mL Multi-Core™ 10X Buffer
Concentration 10U/μL
For Use With (Application) Assayed to ensure optimal performance in genomic analysis applications
Storage Requirements -20°C
Cut Site AAT.ATT TTA.TAA
Format Liquid
Product Name SspI
Quantity 500U
Format Liquid
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
pH 7.4
Quantity 500U
Units 500U
Applications Assayed to ensure optimal performance in genomic analysis applications