Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - SnaB I (B) DFS Item


Manufacturer: promega™  R6791

 View more versions of this product

Catalog No. PR-R6791

  • / Each
 

Description & Specifications

Specifications

Incubator Temperature 37°C
Units 100U
pH 7.3
Product Name SnaBI
Storage Buffer 10mM Tris HCl (pH 7.3), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C
Concentration 2 to 10U/μL
Cut Site TAC.GTA ATG.CAT
Quantity 100U
Format Liquid
Includes 1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 100U SnaBI, 1 x 0.25mL Muti-Core™10X Buffer