missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - SnaB I (B) DFS Item

$125.99 - $491.00

Specifications

Concentration 2 to 10U/μL
Cut Site TAC.GTA ATG.CAT
Incubator Temperature 37°C
pH 7.3
Storage Buffer 10mM Tris HCl (pH 7.3), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Quantity Units Price Quantity  
Catalog Number Mfr. No. Includes Quantity Units Price Quantity  
PR-R6791 Promega™
R6791
1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 100U SnaBI, 1 x 0.25mL Muti-Core™10X Buffer 100U 100U Each for $125.99
PR-R6795 Promega™
R6795
1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U SnaBI, 1 x 0.25mL Muti-Core™10X Buffer 500U 500U Each for $491.00
Specifications

Description

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Description & Specifications

Specifications

2 to 10U/μL
37°C
10mM Tris HCl (pH 7.3), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
TAC.GTA ATG.CAT
7.3
-20°C