Please login to your online account to display your discounted pricing

Restriction Enzymes - Sma I Blue/White Cloning and Genome Qualified (J)


Manufacturer: promega corportaion  R4124

 View more versions of this product

Catalog No. PR-R4124

  • / Each
 

Description & Specifications

Specifications

Quantity 5000U
Product Name SmaI (HC)
Format Liquid
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Concentration 40 to 80U/μL
Includes 1 x 1mL 10X Buffer J, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 5000U SmaI (HC), 1 x 0.25mL Multi-Core™ 10X Buffer
For Use With (Application) Provides a higher level of quality control for Enzymes used in cloning applications
Storage Requirements -20°C
pH 7.4
Cut Site CCC.GGG GGG.CCC
Units 5,000U
Incubator Temperature 37°C